Merck
  • Myoglobin promotes nitrite-dependent mitochondrial S-nitrosation to mediate cytoprotection after hypoxia/reoxygenation.
Myoglobin promotes nitrite-dependent mitochondrial S-nitrosation to mediate cytoprotection after hypoxia/reoxygenation.
Nitric oxide : biology and chemistry (2020-09-07)
Kelly Quesnelle, Danielle A Guimaraes, Krithika Rao, Anuradha Bharara Singh, Yinna Wang, Neil Hogg, Sruti Shiva
ABSTRACT
It is well established that myoglobin supports mitochondrial respiration through the storage and transport of oxygen as well as through the scavenging of nitric oxide. However, during ischemia/reperfusion (I/R), myoglobin and mitochondria both propagate myocardial injury through the production of oxidants. Nitrite, an endogenous signaling molecule and dietary constituent, mediates potent cardioprotection after I/R and this effect relies on its interaction with both myoglobin and mitochondria. While independent mechanistic studies have demonstrated that nitrite-mediated cardioprotection requires the presence of myoglobin and the post-translational S-nitrosation of critical cysteine residues on mitochondrial complex I, it is unclear whether myoglobin directly catalyzes the S-nitrosation of complex I or whether mitochondrial-dependent nitrite reductase activity contributes to S-nitrosation. Herein, using purified myoglobin and isolated mitochondria, we characterize and directly compare the nitrite reductase activities of mitochondria and myoglobin and assess their contribution to mitochondrial S-nitrosation. We demonstrate that myoglobin is a significantly more efficient nitrite reductase than isolated mitochondria. Further, deoxygenated myoglobin catalyzes the nitrite-dependent S-nitrosation of mitochondrial proteins. This reaction is enhanced in the presence of oxidized (Fe3+) myoglobin and not significantly affected by inhibitors of mitochondrial respiration. Using a Chinese Hamster Ovary cell model stably transfected with human myoglobin, we show that both myoglobin and mitochondrial complex I expression are required for nitrite-dependent attenuation of cell death after anoxia/reoxygenation. These data expand the understanding of myoglobin's role both as a nitrite reductase to a mediator of S-nitrosation and as a regulator of mitochondrial function, and have implications for nitrite-mediated cardioprotection after I/R.
Sigma-Aldrich
MISSION® esiRNA, targeting human NDUFAF1
Pricing and availability is not currently available.