Merck
  • Vibrational Dynamics of Crystalline 4-Phenylbenzaldehyde from INS Spectra and Periodic DFT Calculations.
Vibrational Dynamics of Crystalline 4-Phenylbenzaldehyde from INS Spectra and Periodic DFT Calculations.
Molecules (Basel, Switzerland) (2020-03-22)
Mariela M Nolasco, Catarina F Araujo, Pedro D Vaz, Ana M Amado, Paulo Ribeiro-Claro
ABSTRACT
The present work emphasizes the value of periodic density functional theory (DFT) calculations in the assessment of the vibrational spectra of molecular crystals. Periodic calculations provide a nearly one-to-one match between the calculated and observed bands in the inelastic neutron scattering (INS) spectrum of crystalline 4-phenylbenzaldehyde, thus validating their assignment and correcting previous reports based on single molecule calculations. The calculations allow the unambiguous assignment of the phenyl torsional mode at ca. 118-128 cm-1, from which a phenyl torsional barrier of ca. 4000 cm-1 is derived, and the identification of the collective mode involving the antitranslational motion of CH···O bonded pairs, a hallmark vibrational mode of systems where C-H···O contacts are an important feature.
Sigma-Aldrich
Biphenyl-4-carboxaldehyde, 99%
Pricing and availability is not currently available.