Merck
  • Molecular layer interneurons shape the spike activity of cerebellar Purkinje cells.
Molecular layer interneurons shape the spike activity of cerebellar Purkinje cells.
Scientific reports (2019-02-12)
Amanda M Brown, Marife Arancillo, Tao Lin, Daniel R Catt, Joy Zhou, Elizabeth P Lackey, Trace L Stay, Zhongyuan Zuo, Joshua J White, Roy V Sillitoe
ABSTRACT
Purkinje cells receive synaptic input from several classes of interneurons. Here, we address the roles of inhibitory molecular layer interneurons in establishing Purkinje cell function in vivo. Using conditional genetics approaches in mice, we compare how the lack of stellate cell versus basket cell GABAergic neurotransmission sculpts the firing properties of Purkinje cells. We take advantage of an inducible Ascl1CreER allele to spatially and temporally target the deletion of the vesicular GABA transporter, Vgat, in developing neurons. Selective depletion of basket cell GABAergic neurotransmission increases the frequency of Purkinje cell simple spike firing and decreases the frequency of complex spike firing in adult behaving mice. In contrast, lack of stellate cell communication increases the regularity of Purkinje cell simple spike firing while increasing the frequency of complex spike firing. Our data uncover complementary roles for molecular layer interneurons in shaping the rate and pattern of Purkinje cell activity in vivo.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Ankyrin-G Antibody, clone N106/36, clone N106/36, from mouse
Sigma-Aldrich
Anti-Neurogranin Antibody, Chemicon®, from rabbit