
Warfarin Sodium

Synonym(s):
3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
Linear Formula:
C19H15O4Na
CAS Number:
Molecular Weight:
330.31
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2