Skip to Content
Merck
Sodium butyrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sodium butyrate

Sodium butyrate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Butyric acid sodium salt, Sodium Butyrate, Sodium butyrate
Linear Formula:
CH3CH2CH2COONa
CAS Number:
Molecular Weight:
110.09
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique