
Lisinopril R,S,S-diketopiperazine

Synonym(s):
(αS,3S,8aR)-3-(4-Aminobutyl)hexahydro-1,4-dioxo-α-(2-phenylethyl)pyrrolo[1,2-a]pyrazine-2(1H)-acetic acid
Empirical Formula (Hill Notation):
C21H29N3O4
CAS Number:
Molecular Weight:
387.47
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing