Skip to Content
Merck
Guanosine 5′-[β-thio]diphosphate trilithium salt ≥85% (HPLC), powder

Guanosine 5′-[β-thio]diphosphate trilithium salt

Guanosine 5′-[β-thio]diphosphate trilithium salt ≥85% (HPLC), powder
Synonym(s):
GDPβS trilithium salt, GDP-β-S, GDP-β-S-Li3
Empirical Formula (Hill Notation):
C10H12Li3N5O10P2S
CAS Number:
Molecular Weight:
477.07

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing