
Clopamide for system suitability

Synonym(s):
Clopamide, 4-Chloro-N-(2,6-dimethylpiperidinyl)-3-sulfamoylbenzamide
Empirical Formula (Hill Notation):
C14H20ClN3O3S
CAS Number:
Molecular Weight:
345.84
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing