Skip to Content
Merck
β-Nicotinamide adenine dinucleotide phosphate disodium salt ≥97% (calc. based on dry substance, enzymatic)

β-Nicotinamide adenine dinucleotide phosphate disodium salt

β-Nicotinamide adenine dinucleotide phosphate disodium salt ≥97% (calc. based on dry substance, enzymatic)
Synonym(s):
β-NADH phosphate disodium salt, β-NADP-Na2, Triphosphopyridine nucleotide disodium salt
Empirical Formula (Hill Notation):
C21H26N7Na2O17P3
CAS Number:
Molecular Weight:
787.37
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment