
α-Zearalenol

Synonym(s):
2,4-Dihydroxy-6-(6α,10-dihydroxy-trans-1-undecenyl)benzoic acid μ-lactone
Empirical Formula (Hill Notation):
C18H24O5
CAS Number:
Molecular Weight:
320.38
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing