
α-D-Glucose 1,6-bisphosphate tetra(cyclohexylammonium) salt hydrate

Synonym(s):
α-D-Glucose 1,6-diphosphate tetra(cyclohexylammonium) salt hydrate, Glc-1,6-PP.4CHA
Empirical Formula (Hill Notation):
C6H14O12P2 · 4C6H13N · xH2O
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing