WBULP-20ML
Immobilon® ECL UltraPlus Western HRP Substrate
Synonym(s):
HRP substrate, chemiluminescent detection substrate, detection substrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
product name
Immobilon® ECL UltraPlus Western HRP Substrate - 20mL,
product line
Immobilon®
Quality Level
technique(s)
dot blot: suitable
immunoblotting: suitable
western blot (chemiluminescent): suitable
Caution
- 10mL Enhancer Solution
- 10mL Peroxide Solution
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service