Merck
  • Photosynthetic and transcriptional responses of the marine diatom Thalassiosira pseudonana to the combined effect of temperature stress and copper exposure.
Photosynthetic and transcriptional responses of the marine diatom Thalassiosira pseudonana to the combined effect of temperature stress and copper exposure.
Marine pollution bulletin (2017-04-04)
Priscilla T Y Leung, Andy Xianliang Yi, Jack C H Ip, Sarah S T Mak, Kenneth M Y Leung
ABSTRACT
A 96-h exposure experiment was conducted to elucidate the toxicity responses of the marine diatom Thalassiosira pseudonana upon exposure to different temperatures and copper (Cu) concentrations. Three Cu treatments (seawater control; 200μg/L Cu, EC
Sigma-Aldrich
Copper(II) sulfate pentahydrate, JIS special grade, ≥99.5%