Merck
Western Diet Triggers NLRP3-Dependent Innate Immune Reprogramming.
Cell (2018-01-13)
Anette Christ, Patrick Günther, Mario A R Lauterbach, Peter Duewell, Debjani Biswas, Karin Pelka, Claus J Scholz, Marije Oosting, Kristian Haendler, Kevin Baßler, Kathrin Klee, Jonas Schulte-Schrepping, Thomas Ulas, Simone J C F M Moorlag, Vinod Kumar, Min Hi Park, Leo A B Joosten, Laszlo A Groh, Niels P Riksen, Terje Espevik, Andreas Schlitzer, Yang Li, Michael L Fitzgerald, Mihai G Netea, Joachim L Schultze, Eicke Latz
ABSTRACT
Long-term epigenetic reprogramming of innate immune cells in response to microbes, also termed "trained immunity," causes prolonged altered cellular functionality to protect from secondary infections. Here, we investigated whether sterile triggers of inflammation induce trained immunity and thereby influence innate immune responses. Western diet (WD) feeding of Ldlr
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)