Merck
  • Introduction of antisense oligonucleotides into cells by permeabilization with streptolysin O.
Introduction of antisense oligonucleotides into cells by permeabilization with streptolysin O.
BioTechniques (1993-12-01)
E L Barry, F A Gesek, P A Friedman
ABSTRACT
Cellular uptake of antisense oligonucleotides is critical to their ability to inhibit gene expression. In the present study, phosphodiester oligodeoxynucleotides were introduced into cells during brief permeabilization with the pore-forming agent streptolysin O. The extent of antisense inhibition was dependent on the concentration of oligonucleotide present during permeabilization. In addition, the level of antisense inhibition was time-dependent; it reached a maximum at 18 h and subsequently diminished to control levels over the next 48 h. These results demonstrate the effectiveness of streptolysin O permeabilization as a means for simple and rapid introduction of oligonucleotides into eukaryotic cells.
Sigma-Aldrich
Streptolysin O from Streptococcus pyogenes, ≥1,000,000 units/mg protein, recombinant, lyophilized powder, expressed in E. coli
Pricing and availability is not currently available.