Merck
  • The solubilisation of the membrane-bound D-alanyl-D-alanine carboxypeptidase of Bacillus coagulans NCIB 9365.
The solubilisation of the membrane-bound D-alanyl-D-alanine carboxypeptidase of Bacillus coagulans NCIB 9365.
Biochimica et biophysica acta (1979-06-06)
H A McArthur, P E Reynolds
PMID39605
ABSTRACT
Protoplast membranes and the particulate D,D-carboxypeptidase of Bacillus coagulans NCIB 9365 were extremely resistant to disruption by either detergents or urea. A combination of urea and the non-ionic detergent Genapol X-100 was required to achieve a significant solubilisation of membrane protein and D,D-carboxypeptidase in an active form; the pH optimum for this treatment was pH 7.5. Solubilisation of the enzyme was accompanied by a two-fold enhancement of activity. Kinetic results indicated that the enhancement may be due to an alteration in the conformation of the enzyme following disruption of membrane structure.
Sigma-Aldrich
Genapol® C-100
Sigma-Aldrich
Genapol® X-100