Merck
  • In vivo imaging of therapy response to a novel pan-HER antibody mixture using FDG and FLT positron emission tomography.
In vivo imaging of therapy response to a novel pan-HER antibody mixture using FDG and FLT positron emission tomography.
Oncotarget (2015-10-16)
Carsten H Nielsen, Mette M Jensen, Lotte K Kristensen, Anna Dahlman, Camilla Fröhlich, Helle J Jacobsen, Thomas T Poulsen, Johan Lantto, Ivan D Horak, Michael Kragh, Andreas Kjaer
ABSTRACT
Overexpression of the human epidermal growth factor receptor (HER) family and their ligands plays an important role in many cancers. Targeting multiple members of the HER family simultaneously may increase the therapeutic efficacy. Here, we report the ability to image the therapeutic response obtained by targeting HER family members individually or simultaneously using the novel monoclonal antibody (mAb) mixture Pan-HER. Mice with subcutaneous BxPC-3 pancreatic adenocarcinomas were divided into five groups receiving vehicle or mAb mixtures directed against either EGFR (HER1), HER2, HER3 or all three receptors combined by Pan-HER. Small animal positron emission tomography/computed tomography (PET/CT) with 2'-deoxy-2'-[(18)F]fluoro-D-glucose (FDG) and 3'-deoxy-3'-[(18)F]fluorothymidine (FLT) was performed at baseline and at day 1 or 2 after initiation of therapy. Changes in tumor uptake of tracers were quantified and compared to reduction in tumor size. Imaging results were further validated by immunohistochemistry and qPCR. Mean FDG and FLT uptake in the Pan-HER treated group decreased by 19 ± 4.3% and 24 ± 3.1%, respectively. The early change in FDG and FLT uptake correlated with tumor growth at day 23 relative to day 0. Ex vivo molecular analyses of markers associated with the mechanisms of FDG and FLT uptake confirmed the in vivo imaging results. Taken together, the study supports the use of FDG and FLT as imaging biomarkers of early response to Pan-HER therapy. FDG and FLT PET/CT imaging should be considered as imaging biomarkers in clinical evaluation of the Pan-HER mAb mixture.
Sigma-Aldrich
Anti-HK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-1138981-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€414.20
EN300-1138981-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€571.50
EN300-1138981-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€871.80
EN300-1138981-1G1 g
Ships in 14 days - from Aldrich Partner.
€1,105.80
EN300-1138981-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
€2,123.70
EN300-1138981-5G5 g
Ships in 14 days - from Aldrich Partner.
€3,120.80
EN300-1138981-10G10 g
Ships in 14 days - from Aldrich Partner.
€4,605.40
To order products, please contact your local dealer.