Merck
  • Comparative intestinal esterases amongst passerine species: Assessing vulnerability to toxic chemicals in a phylogenetically explicit context.
Comparative intestinal esterases amongst passerine species: Assessing vulnerability to toxic chemicals in a phylogenetically explicit context.
Chemosphere (2015-04-29)
Cristobal Narvaez, Natalia Ramirez-Otarola, Francisco Bozinovic, Juan C Sanchez-Hernandez, Pablo Sabat
ABSTRACT
Inhibition of blood esterase activities by organophosphate (OP) pesticides has been used as a sensitive biomarker in birds. Furthermore, compared to mammalian vertebrates, less is known about the role of these enzyme activities in the digestive tracts of non-mammalian vertebrates, as well as the environmental and biological stressors that contribute to their natural variation. To fill this gap, we examined butyrylcholinesterase (BChE) and carboxylesterases (CbE) in the digestive tracts of sixteen passerine species from central Chile. Whole intestine enzyme activities were positively and significantly correlated with body mass. After correcting for body mass and phylogenetic effect, we found only a marginal effect of dietary category on BChE activity, but a positive and significant association between the percentage of dietary nitrogen and the mass-corrected lipase activity. Our results suggest that observed differences may be due to the dietary composition in the case of lipases and BChE, and also we predict that all model species belonging to the same order will probably respond differently to pesticide exposure, in light of differences in the activity levels of esterase activities.
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate
Sigma-Aldrich
Butyrylthiocholine iodide, ≥98%