Merck
  • Biotransformation of 1,3-propanediol cyclic sulfate and its derivatives to diols by toluene-permeabilized cells of Bacillus sp. CCZU11-1.
Biotransformation of 1,3-propanediol cyclic sulfate and its derivatives to diols by toluene-permeabilized cells of Bacillus sp. CCZU11-1.
Applied biochemistry and biotechnology (2014-12-31)
Yu-Cai He, Feng Liu, Dan-Ping Zhang, Shan Gao, Zhi-Qing Li, Zhi-Cheng Tao, Cui-Luan Ma
ABSTRACT
In this study, it was the first report that Bacillus sp. CCZU11-1 was used for the biotransformation of 1,3-propanediol cyclic sulfate (1,3-PDS) and its derivatives. The catalytic performance of Bacillus sp. sulfatase in the biotransformation of 1,3-PDS was significantly improved by biocatalyst permeabilization and immobilization. Using cell permeabilization, the hydrolytic activity of the whole-cell biocatalyst was increased by 3.5-fold after 1.5 h of pretreatment with 10 % (v/v) toluene at 30 °C and pH 7.0. Biotransformation of 20 mM 1,3-PDS for 24 h, 1,3-propanediol (1,3-PD) could be obtained in the yield of 97.4 % under the optimized reaction condition. Additionally, the immobilized biocatalysts, permeabilized cells entrapped in calcium alginate, and cross-linked enzyme aggregates were further employed to biotansform 1,3-PDS. Moreover, the total operational time of the immobilized biocatalysts could reach above 240 h with high conversion rate (>90 %).
Sigma-Aldrich
1,3-Propanediol cyclic sulfate, 98%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%