Merck
  • Neutrophil infiltration during inflammation is regulated by PILRα via modulation of integrin activation.
Neutrophil infiltration during inflammation is regulated by PILRα via modulation of integrin activation.
Nature immunology (2012-11-13)
Jing Wang, Ikuo Shiratori, Junji Uehori, Masahito Ikawa, Hisashi Arase
ABSTRACT
Acute inflammatory responses are important in host defense, whereas dysregulated inflammation results in life-threatening complications. Here we found that paired immunoglobulin-like type 2 receptor alpha (PILRα), an inhibitory receptor containing immunoreceptor tyrosine-based inhibitory motifs (ITIMs), negatively regulated neutrophil infiltration during inflammation. Pilra(-/-) mice had increased neutrophil recruitment to inflammatory sites and were highly susceptible to endotoxin shock. Pilra(-/-) neutrophils showed enhanced transmigration ability and increased adhesion to the β(2) integrin ligand ICAM-1. PILRα expressed on neutrophils constitutively associated in cis with its ligands, resulting in clustering of PILRα during stimulation with a chemoattractant. Clustering of PILRα enhanced ITIM-mediated signaling, thus modulating β(2) integrin inside-out activation. These data demonstrate that neutrophil recruitment in inflammatory responses is regulated by PILRα via modulation of integrin activation.
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)