Merck
  • Determination of thiodicarb using a biosensor based on alfalfa sprout peroxidase immobilized in self-assembled monolayers.
Determination of thiodicarb using a biosensor based on alfalfa sprout peroxidase immobilized in self-assembled monolayers.
Talanta (2010-08-06)
Sally K Moccelini, Iolanda C Vieira, Fábio de Lima, Bruno G Lucca, Antônio M J Barbosa, Valdir S Ferreira
ABSTRACT
A biosensor based on alfalfa sprout (Medicago sativa) homogenate as a source of peroxidase is proposed for the determination of thiodicarb by square-wave voltammetry. This enzyme was immobilized in self-assembled monolayers of l-cysteine on a gold electrode. Several parameters were investigated to evaluate the optimum conditions for operation of the biosensor. The analytical curve was linear for thiodicarb concentrations of 2.27 x 10(-6) to 4.40 x 10(-5) mol L(-1) with a detection limit of 5.75 x 10(-7) mol L(-1). The lifetime of the Au-alfalfa sprout-SAMs was 20 days (at least 220 determinations). The average recovery of thiodicarb from samples of vegetable extracts ranged from 99.02 to 101.04%. The results obtained for thiodicarb in vegetable extracts using the proposed method are in close agreement with those using a high performance liquid chromatography procedure at the 95% confidence level.
Product Description
Supelco
Thiodicarb, PESTANAL®, analytical standard
Pricing and availability is not currently available. Contact Technical Service