Merck
  • Quantitative determination of TEGDMA, BHT, and DMABEE in eluates from polymerized resin-based dental restorative materials by use of GC/MS.
Quantitative determination of TEGDMA, BHT, and DMABEE in eluates from polymerized resin-based dental restorative materials by use of GC/MS.
Archives of toxicology (2009-09-23)
Mario Seiss, Christopher Langer, Reinhard Hickel, Franz-Xaver Reichl
ABSTRACT
This study investigated the leaching of ingredients from several commercial dental composite resins cured with LED, and immersed in methanol or water for 24 h, respectively. The composites used were: Admira Dentin (VOCO), Artemis Schmelz (Enamel) (Ivoclar Vivadent), Els extra low shrinkage (Saremco Dental), Filtek Supreme XT Dentin (3 M ESPE), Gradia Direct (GC), Venus & Venus flow (Heraeus Kulzer), and XRV Herculite Prodigy Enamel (Kerr). From each dental composite four specimens with defined structure and 100-mg net weight were made. After the polymerization process, according to manufacturer's instructions, the specimens were immersed in either 1 ml water or 1 ml methanol and incubated at 37 degrees C for 24 h. Eluted ingredients triethyleneglycoldimethacrylate (TEGDMA), 2,6-di-tert-butyl-4-methylphenol (BHT), and 4-N,N-dimethylaminobenzoicacidethylester (DMABEE) were detected and quantified using gas chromatography-mass spectrometry (GC-MS). The amounts of the detected analytes from 100 mg polymerized composites ranged between the following values: TEGDMA: 0-0.5 mg (water), 0-1.6 mg (methanol); BHT: 0-0.03 μg (water), 0-0.11 mg (methanol); and DMABEE: 0-0.11 mg (water), 0-1.4 mg (methanol). We conclude from the results that the elution rates into methanol and water differ significantly. Furthermore, it is concluded that all the determined amounts eluting from the composites are far below toxic-relevant concentrations.
Product Description
Supelco
Ethyl 4-(dimethylamino)benzoate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethyl 4-(dimethylamino)benzoate, ≥99%