Merck
  • Acrolein sequestering ability of the endogenous tripeptide glycyl-histidyl-lysine (GHK): characterization of conjugation products by ESI-MSn and theoretical calculations.
Acrolein sequestering ability of the endogenous tripeptide glycyl-histidyl-lysine (GHK): characterization of conjugation products by ESI-MSn and theoretical calculations.
Journal of pharmaceutical and biomedical analysis (2008-04-02)
Giangiacomo Beretta, Emanuele Arlandini, Roberto Artali, Josep M Garcia Anton, R Maffei Facino
ABSTRACT
Acrolein (ACR) is a well-known carbonyl toxin produced by lipid peroxidation of polyunsaturated fatty acids, which is involved in several life-threatening pathologies such as Alzheimer disease, arteriosclerosis, diabetes, and nephropathy. The aim of this work was to study the quenching ability of the endogenous tripeptide glycyl-histidyl-lysine (GHK), a liver cell growth factor isolated from human plasma, towards the electrophilic aldehyde ACR and to characterize the reaction products by electrospray mass spectrometry (ESI-MS/MS infusion experiments; positive ion mode). The reaction of ACR (30 microM) with GHK (0.1, 0.25, 0.5, 1.0 mM) was followed by measuring aldehyde consumption by reverse-phase HPLC (phosphate buffer, pH 7.4); after 4h, when the aldehyde had completely disappeared; the reaction products were checked by ESI-MS/MS. Several products were detected in the GHK+ACR reaction (1:1). This indicates a complex reaction cascade involving the sequential addition of ACR (up to 3 mol) to the tripeptide GHK and, in particular, to the epsilon-amino group of the lysine residue and to the N(tau) and N(pi) of the histidine moiety. The Michael addition of two molecules of ACR to the epsilon-amino group of the lysine residue is followed by aldol condensation and dehydration to give the N-(3-formyl-3,4-dehydropiperidino) derivative. The results confirm that the ESI-MS/MS approach in a direct infusion experiment permits rapid profiling of the products of the GHK+ACR reaction. They firstly point to the potential medicinal use of GHK in the prevention of carbonyl stress-linked pathologies, and--second--help shed light on the physiological role of this histidine-containing tripeptide which is claimed to be an endogenous growth factor, but has never been shown to be an ACR quencher.
Sigma-Aldrich
Gly-His-Lys acetate salt, ≥97% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture
Pricing and availability is not currently available.