Merck
  • Wntless expression promotes lineage plasticity and is associated with neuroendocrine prostate cancer.
Wntless expression promotes lineage plasticity and is associated with neuroendocrine prostate cancer.
American journal of clinical and experimental urology (2022-11-01)
Leandro S D'Abronzo, Alan P Lombard, Shu Ning, Cameron M Armstong, Amy R Leslie, Masuda Sharifi, Zachary A Schaaf, Wei Lou, Allen C Gao
ABSTRACT
Resistance to androgen receptor (AR) targeted therapies remains as the main reason for most prostate cancer related deaths. Lineage plasticity resulting in altered, treatment insensitive prostate tumor cell phenotypes such neuroendocrine differentiated prostate cancer is a common manifestation within resistant tumors upon AR-targeted therapies. The mechanisms responsible for lineage plasticity in prostate cancer remain incompletely understood. Here we demonstrate that the enzalutamide resistant MDVR cell line possesses lineage plastic characteristics associated with overexpression of the Wnt transporter Wntless (WLS). Furthermore, we present evidence that overexpression of WLS is common in varying cell line models of lineage plastic prostate cancer, is higher in neuroendocrine patient samples, and positively correlates with the neuroendocrine marker SYP in clinical data. Targeting WLS in lineage plastic cellular models reduces viability and represses lineage plasticity associated gene expression. Our study provides insight into the importance of WLS to the development of lethal resistant prostate cancer and provides a potential target for the treatment of advanced disease.
Sigma-Aldrich
Anti-GPR177 Antibody, clone YJ5, clone YJ5, from mouse
Sigma-Aldrich
Anti-Neuron Specific Enolase Antibody, clone 5E2, clone 5E2, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2