Merck
  • miR-141 regulates KEAP1 and modulates cisplatin sensitivity in ovarian cancer cells.
miR-141 regulates KEAP1 and modulates cisplatin sensitivity in ovarian cancer cells.
Oncogene (2012-10-10)
M T M van Jaarsveld, J Helleman, A W M Boersma, P F van Kuijk, W F van Ijcken, E Despierre, I Vergote, R H J Mathijssen, E M J J Berns, J Verweij, J Pothof, E A C Wiemer
ABSTRACT
Epithelial ovarian cancer is the most lethal gynecological malignancy in the Western world. A major impediment for the successful treatment is the development of drug resistance. The molecular processes that contribute to resistance have been extensively studied; however, there is not much known about regulation by microRNAs (miRNAs). We compared miRNA expression profiles of an isogenic cisplatin-sensitive and -resistant ovarian cancer cell line pair (A2780/A2780 DDP) and found 27 miRNAs to be differentially expressed (2-fold). Five of these, including the family members miR-141/200c, showed a correlation with cisplatin sensitivity in the NCI-60 panel. Overexpression of miR-141 resulted in enhanced resistance to cisplatin in ovarian cancer cell lines. We next correlated the expression level of miR-141 in 132 primary ovarian tumors (108 serous and 24 non-serous) with response to platinum-based chemotherapy. Although no differences were observed in the serous tumors, miR-141 levels were higher in non-serous ovarian tumors that did not respond well to therapy (platinum-free interval <6 months). We demonstrate that miR-141 directly targets KEAP1, and that downregulation of KEAP1 induces cisplatin resistance. Conversely, overexpression of KEAP1 significantly enhanced cisplatin sensitivity. Expression of KEAP1 with its 3'-UTR, and a 3'-UTR in which the miR-141 target site has been mutated, revealed that miR-141 regulates KEAP1 upon exposure to cisplatin. Finally, we show that the NF-κB pathway, which can be regulated by KEAP1, is activated upon miR-141 overexpression, and that inhibition of this pathway partially reverses miR-141-mediated cisplatin resistance. These findings demonstrate that the miR-141-mediated regulation of KEAP1 has a crucial role in the cellular response to cisplatin.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-MIB1 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution