Merck
  • Loading, stationary phase, and salt effects during hydrophobic interaction chromatography: alpha-lactalbumin is stabilized at high loadings.
Loading, stationary phase, and salt effects during hydrophobic interaction chromatography: alpha-lactalbumin is stabilized at high loadings.
Journal of chromatography. A (2006-05-13)
Jace L Fogle, John P O'Connell, Erik J Fernandez
ABSTRACT
Amide hydrogen-deuterium exchange labeling has been used to study the effects of salt and protein loading on alpha-lactalbumin (BLA) stability during hydrophobic interaction chromatography (HIC). Stability in the adsorbed phase increased dramatically with increasing loading, and unfolding was nearly undetectable close to the resin saturation capacity. We also found that a butyl surface destabilized BLA more than a phenyl surface, despite the fact that BLA was bound more strongly on the phenyl surface. These observations have important implications for HIC process design and indicate that in some cases column capacity does not have to be sacrificed to preserve protein stability.
Sigma-Aldrich
Phenyl-Sepharose 6 Fast Flow, low substitution, extent of labeling: ~20 μmol per mL
Sigma-Aldrich
Phenyl-Sepharose 6 Fast Flow, high substitution, extent of labeling: ~40 μmol per mL (high substitution)