Merck
  • Comparative antioxidant and antidiabetic effects of Syzygium polyanthumleaf and Momordica charantia fruit extracts.
Comparative antioxidant and antidiabetic effects of Syzygium polyanthumleaf and Momordica charantia fruit extracts.
Pakistan journal of pharmaceutical sciences (2018-04-08)
Muhammad Jihad Sandikapura, Shaik Nyamathulla, Mohamed Ibrahim Noordin
ABSTRACT
The study was aimed to perform aqueous extraction of two plants using different extraction methods, and evaluate their antioxidant and antidiabetic potential. Plant materials were extracted by maceration, soxhlet, sonication and fresh juice methods to produce aqueous extracts. In vitro antioxidant DPPH (1,1-diphenyl-2- picrylhydrazyl) and FRAP (Ferric reducing antioxidant power), antidiabetic α-amylase and α-glucosidase enzyme inhibitory assays were carried out on the extracts. Extracts of Syzygium polyanthum demonstrated better free radical scavenging and antidiabetic activity than Momordica charantia. It was observed that the % inhibition of DPPH by fresh juice of S. polyanthum was 64.93 similar to quercetin 69.21 (p>0.05). Its FRAP value (69.05) was significantly (p<0.05) higher than Quercetin (63.27). Its fresh juice alsodemonstrated significant inhibitory actions (p<0.05) against α-amylase (92.21%) and α-glucosidase (96.06%) than acarbose. It is concluded that extracts had varied results due to differences in their chemical composition as noticed in LC-MS. The fresh juice of S. polyanthum has superior in vitro antioxidant and antidiabetic activities. Therefore, intake of exogenous antioxidants in the form of fresh juices of someherbs can help the body toscavenge free radicals and exert hyperglycaemic control in post prandial hyperglycaemia.
Sigma-Aldrich
Acarbose, ≥95% (HPLC)