Skip to Content
Merck
vibrant-m

Oxaliplatin Related Compound C

vibrant-m
Synonym(s):
(OC-6-33)-[(1R,2R)-Cyclohexane-1,2-diamine-κNN′][ethanedioato(2-)-κO1O2]dihydroxyplatinum
Empirical Formula (Hill Notation):
C8H16N2O6Pt
CAS Number:
Molecular Weight:
431.30

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1