
Nuclear fast red (C.I. 60760)

Synonym(s):
Nuclear fast red (C.I. 60760)
Empirical Formula (Hill Notation):
C14H8NNaO7S
CAS Number:
Molecular Weight:
357.27
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing