Skip to Content
Merck
Monoclonal Anti-c-Myc antibody produced in mouse clone 9E10, ascites fluid

Monoclonal Anti-c-Myc antibody produced in mouse

Monoclonal Anti-c-Myc antibody produced in mouse clone 9E10, ascites fluid
Synonym(s):
Anti-c-Myc

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing