
DL-Glyceric acid hemicalcium salt hydrate

Synonym(s):
2,3-Dihydroxypropanoic acid hemicalcium salt, Hemicalcium DL-glycerate, hemi-Calcium DL-glycerate
Linear Formula:
[HOCH2CH(OH)COO]2Ca · ×H2O
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing