
Codeine sulfate

Synonym(s):
Codeine sulfate salt, 3-Methylmorphine sulfate salt
Linear Formula:
C36H42N2O6 · H2SO4
CAS Number:
Molecular Weight:
696.81
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing