
Anti-Actin (plant) antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-Actin (plant) antibody produced in mouse
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing