
6-Hexachloro-Fluorescein Phosphoramidite

Synonym(s):
6-(4,7,2′,4′,5′,7′-Hexachloro-3′,6′-dipivaloylfluoresceinyl-6-carboxamido)-hexyl-1-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
Empirical Formula (Hill Notation):
C46H52Cl6N3O10P
CAS Number:
Molecular Weight:
1050.61
Products
Product Category
Brand
Color
Formula Weight
Purity
Quality Segment
Showing 1-3 of 3