
3-(Boc-amino)propyl bromide

Synonym(s):
tert-Butyl N-(3-bromopropyl)carbamate
Linear Formula:
Br(CH2)3NHCO2C(CH3)3
CAS Number:
Molecular Weight:
238.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing