
2,4-Dichloro-5-sulfamoylbenzoic acid

Linear Formula:
Cl2C6H2(SO2NH2)CO2H
CAS Number:
Molecular Weight:
270.09
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing