The samples were prepared by dissolving approximately 10 mg of PHR2570 in 100 mL of diluent, which was prepared as follows:
1L water, 1L acetonitrile, and 1mL concentrated phosphoric acid.
PHR2570
Riboflavin Impurity B
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
7,8-dimethylbenzo[g]pteridine-2,4(1H,3H)-dione
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
riboflavin
form
solid
packaging
pkg of 50 mg
application(s)
pharmaceutical
storage temp.
2-8°C
General description
Application
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Could you provide details about the HPLC sample preparation procedure for PHR2570? A customer tried methanol 0.2 mg/ml but did not succeed in solubilizing the product.
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service