Merck
HomeCell SignalingUse of the Protease Fluorescent Detection Kit to Determine Protease Activity
Use of the Protease Fluorescent Detection Kit to Determine Protease Activity
The Protease Fluorescent Detection Kit provides ready-to-use reagents for detecting the presence of protease activity. This simple assay to detect protease activity uses casein labeled with fluorescein isothiocyanate (FITC) as the substrate.
Protease activity results in the cleavage of the FITC-labeled casein substrate into smaller fragments, which do not precipitate under acidic conditions. After incubation of the protease sample and substrate, the reaction is acidified with the addition of trichloroacetic acid (TCA). The mixture is then centrifuged with the undigested substrate forming a pellet and the smaller, acid soluble fragments remaining in solution. The supernatant is neutralized and the fluorescence of the FITC-labeled fragments is measured.
The described kit procedure detects the trypsin protease control at a concentration of approximately 0.5 μg/mL (5 ng of trypsin added to the assay). This sensitivity can be increased with a longer incubation time, up to 24 hours. The assay is performed in microcentrifuge tubes and procedures are provided for fluorescence detection using either cuvettes or multiwell plates.
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Cupp-Enyard C. Use of the Protease Fluorescent Detection Kit to Determine Protease Activity. JoVE.(30): https://doi.org/10.3791/1514