HPLC Analysis of HSA Fusion Protein on Proteomix® SAX-NP5
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
pressure
2016 psi (139 bar)
column temp.
30 °C
column
Proteomix SAX-NP5 25 cm x 4.6mm I.D., 5 μm particles (Z777229)
detector
UV, 280 nm
flow rate
0.5 mL/min
mobile phase
Piperazine, Triethanolamine, bis-tris propane and N-methylpiperazine, 20 mM each,(A) pH 9.7; (B) pH 3.0 ( adjust by HCl )
sample
HSA fusion peptide 5 mg/mL, 10 μL (MW 75 kD, pI 5.0, HSA conjugated peptide in diabetes treatment)
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies