Merck
  • Structural and functional insights into the lipopolysaccharide ABC transporter LptB
Structural and functional insights into the lipopolysaccharide ABC transporter LptB
Nature communications (2017-08-10)
Haohao Dong, Zhengyu Zhang, Xiaodi Tang, Neil G Paterson, Changjiang Dong
ABSTRACT
The cell surface of most Gram-negative bacteria contains lipopolysaccharide that is essential for their viability and drug resistance. A 134-kDa protein complex LptB
Sigma-Aldrich
Anti-c-Myc−Alkaline Phosphatase antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)