Merck
  • Identification of a histone acetyltransferase as a novel regulator of Drosophila intestinal stem cells.
Identification of a histone acetyltransferase as a novel regulator of Drosophila intestinal stem cells.
FEBS letters (2013-03-29)
Yansen Ma, Zhi Chen, Yaping Jin, Wei Liu
ABSTRACT
One of the major challenges in stem cell research is to decipher the controlling mechanisms/genes of stem cell homeostasis. Through an RNAi mediated genetic screen of living animals, we identified an evolutionarily conserved histone acetyltransferase Atac2 as a novel regulator of Drosophila intestinal stem cells (ISCs). Expression of Atac2-RNAi or a dominant negative allele of Atac2 generated more ISCs, while excessive Atac2 or a histone deacetylase inhibitor promoted ISC differentiation without affecting ISC survival or lineage specification. These findings extend our knowledge of epigenetic mechanisms in stem cell regulation.
Product Description
Roche
Anti-Digoxigenin-Rhodamine, Fab fragments, from sheep
Roche
DIG RNA Labeling Kit (SP6/T7), sufficient for 2 x 10 labeling reactions, kit of 1 (12 components), suitable for hybridization, suitable for Southern blotting