Merck
  • Pd-catalyzed carbonylative lactamization: a novel synthetic approach to FR900482.
Pd-catalyzed carbonylative lactamization: a novel synthetic approach to FR900482.
Organic letters (2004-05-21)
Barry M Trost, Michael K Ameriks
ABSTRACT
An asymmetric synthesis of the benzazocine core of FR900482 has been achieved in 15 steps from 3,5-dinitro-p-toluic acid. Key features of the synthesis include an enantioselective N-methylephedrine-mediated zinc acetylide addition to a highly enolizable arylacetaldehyde and a novel Pd-catalyzed carbonylative lactamization to form an eight-membered ring. [reaction--see text]
Sigma-Aldrich
4-Methyl-3,5-dinitrobenzoic acid, 98%
Sigma-Aldrich
[1,1′-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), complex with dichloromethane