Merck
  • Cyclosporin A specifically inhibits function of nuclear proteins involved in T cell activation.
Cyclosporin A specifically inhibits function of nuclear proteins involved in T cell activation.
Science (New York, N.Y.) (1989-12-22)
E A Emmel, C L Verweij, D B Durand, K M Higgins, E Lacy, G R Crabtree
ABSTRACT
One action of cyclosporin A thought to be central to many of its immunosuppressive effects is its ability to inhibit the early events of T lymphocyte activation such as lymphokine gene transcription in response to signals initiated at the antigen receptor. Cyclosporin A was found to specifically inhibit the appearance of DNA binding activity of NF-AT, AP-3, and to a lesser extent NF-kappa B, nuclear proteins that appear to be important in the transcriptional activation of the genes for interleukin-2 and its receptor, as well as several other lymphokines. In addition, cyclosporin A abolished the ability of the NF-AT binding site to activate a linked promoter in transfected mitogen-stimulated T lymphocytes and in lymphocytes from transgenic mice. These results indicate that cyclosporin A either directly inhibits the function of nuclear proteins critical to T lymphocyte activation or inhibits the action of a more proximal member of the signal transmission cascade leading from the antigen receptor to the nucleus.
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, for molecular biology, ≥95%