Merck
  • Acute thyrotoxicosis secondary to destructive thyroiditis associated with cardiac catheterization contrast dye.
Acute thyrotoxicosis secondary to destructive thyroiditis associated with cardiac catheterization contrast dye.
Thyroid : official journal of the American Thyroid Association (2011-03-10)
Laura Calvi, Gilbert H Daniels
ABSTRACT
Thyrotoxicosis caused by destructive thyroiditis is self-limited and results from the subacute release of preformed thyroid hormone. Common etiologies include painful subacute thyroiditis and silent (painless) subacute thyroiditis (including postpartum thyroiditis, amiodarone-associated destructive thyroiditis, and lithium-associated thyroiditis). Thyrotoxicosis commonly evolves slowly over a matter of weeks. We report a unique case of severe thyrotoxicosis caused by acute- onset painful destructive thyroiditis in a patient who received large amounts of nonionic contrast dye Hexabrix® for cardiac catheterization. The results of thyroid function and physical examination were normal before the catheterization. The acute onset of severe thyroid pain, rapid increase in serum Free Thyroxine Index, and thyroglobulin concentrations with a triiodothyronine to free thyroxine index ratio of < 20 to 1 were compatible with an acute onset destructive thyroiditis, likely related to direct toxicity from the iodinated contrast material. In light of the large number of patients who receive these contrast agents during cardiac catheterization, clinicians should be advised of this potentially serious complication, particularly in the setting of unstable cardiac disease.
Product Description
USP
Ioxaglic acid, United States Pharmacopeia (USP) Reference Standard
Ioxaglic acid, European Pharmacopoeia (EP) Reference Standard