Merck
Caffeine potentiation of propane sultone mutagenesis in barley.
Mutation research (1985-12-01)
C Singh, B L Kaul
ABSTRACT
Pre- and post-treatments with caffeine enhanced propane sultone (PS)-induced growth damage in barley. The caffeine post-treatments were, however, more effective in potentiating PS-induced growth damage and caused an additive effect on the frequency of chlorophyll mutations.
Sigma-Aldrich
1,3-Propanesultone, 98%
Sigma-Aldrich
1,3-Propanesultone, ≥99%