Merck
  • Accumulation of organochlorine pesticides and polychlorinated biphenyls in sediments, aquatic organisms, birds, bird eggs and bat collected from south India.
Accumulation of organochlorine pesticides and polychlorinated biphenyls in sediments, aquatic organisms, birds, bird eggs and bat collected from south India.
Environmental science and pollution research international (2001-05-22)
K Senthilkumar, K Kannan, A Subramanian, S Tanabe
ABSTRACT
Concentrations of polychlorinated biphenyls (PCBs), DDT and its metabolites (DDTs), HCH isomers (HCHs), chlordane compounds (CHLs) and hexachlorobenzene (HCB) were determined in sediment, soil, whole body homogenates of resident and migratory birds and their prey items (including fish, green mussel, snail, earthworm, crabs, prawn, lizard and frogs), bird eggs and bats collected from southern India during 1995 and 1998. Accumulation pattern of organochlorines (OCs) in biota was, in general, in the order, HCHs > DDTs > PCBs > CHLs = HCB. Magnitude of OC concentrations increased in the order of sediments < green mussel < earthworm < from < lizard < fish < bird egg < bats < birds tissues. Biomagnification features of OCs were examined in resident and migrant birds to evaluate the exposure levels of these chemicals in wintering grounds of migrant birds. Accumulation of DDTs in migratory birds during wintering in India may be of concern due to the great biomagnification potential of DDTs. Eggs of some resident species contained noticeable concentrations of OCs. Concentrations of OCs in three species of bats analyzed in this study were lower than that found in passerine birds. In addition to OCs, butyltin compounds were also detected at low concentrations in bats.
Sigma-Aldrich
Butyltin trichloride, 95%