Merck
  • Effect of Water pH on the Uptake of Acidic (Ibuprofen) and Basic (Propranolol) Drugs in a Fish Gill Cell Culture Model.
Effect of Water pH on the Uptake of Acidic (Ibuprofen) and Basic (Propranolol) Drugs in a Fish Gill Cell Culture Model.
Environmental science & technology (2021-03-17)
Elisabeth Dohmann Chang, Raewyn M Town, Stewart F Owen, Christer Hogstrand, Nic R Bury
ABSTRACT
Water pH is predicted to affect the uptake of ionizable pharmaceuticals in fish. The current study used an in vitro primary fish gill cell culture system to assess the effect of pH values in the range of 4.5-8.75 on the uptake rates of the base propranolol (pKa 9.42) and the acid ibuprofen (pKa 4.59). The rate-limiting step in the uptake was the diffusive supply flux of the unionized form from the water to the apical membrane, with subsequent rapid transfer across the epithelium. Computed uptake rate based on the unionized fraction best described the uptake of propranolol and ibuprofen over the range of pH values 5-8 and 6-8.75, respectively. For ibuprofen, the computed uptake rate overestimated the uptake below pH 6 where the unionized fraction increased from 4% at pH 6 to 55% at pH 4.5. As the unionized fraction increased, the uptake rate plateaued suggesting a saturation of the transport process. For both drugs, large variations in the uptake occur with only small fluctuations in pH values. This occurs between pH values 6 and 8, which is the pH range acceptable in regulatory test guidelines and seen in most of our freshwaters.
Sigma-Aldrich
BIS-TRIS propane, ≥99.0% (titration)
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture