Merck
  • Mitochondrial ROS control neuronal excitability and cell fate in frontotemporal dementia.
Mitochondrial ROS control neuronal excitability and cell fate in frontotemporal dementia.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2021-06-01)
Noemí Esteras, Olga Kopach, Marta Maiolino, Vincenzo Lariccia, Salvatore Amoroso, Seema Qamar, Selina Wray, Dmitri A Rusakov, Morana Jaganjac, Andrey Y Abramov
ABSTRACT
The second most common form of early-onset dementia-frontotemporal dementia (FTD)-is often characterized by the aggregation of the microtubule-associated protein tau. Here we studied the mechanism of tau-induced neuronal dysfunction in neurons with the FTD-related 10+16 MAPT mutation. Live imaging, electrophysiology, and redox proteomics were used in 10+16 induced pluripotent stem cell-derived neurons and a model of tau spreading in primary cultures. Overproduction of mitochondrial reactive oxygen species (ROS) in 10+16 neurons alters the trafficking of specific glutamate receptor subunits via redox regulation. Increased surface expression of α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) and N-methyl-D-aspartate (NMDA) receptors containing GluA1 and NR2B subunits leads to impaired glutamatergic signaling, calcium overload, and excitotoxicity. Mitochondrial antioxidants restore the altered response and prevent neuronal death. Importantly, extracellular 4R tau induces the same pathological response in healthy neurons, thus proposing a mechanism for disease propagation. These results demonstrate mitochondrial ROS modulate glutamatergic signaling in FTD, and suggest a new therapeutic strategy.
Sigma-Aldrich
Anti-NMDAR1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-pan-AMPA receptor (GluR1-4), clone 2D8 Antibody, clone 2D8, from mouse