Skip to Content
Merck
vibrant-m

Tolterodine for system suitability

vibrant-m
Synonym(s):
(R)-Tolterodine L-tartrate
Empirical Formula (Hill Notation):
C22H31NO · C4H6O6
CAS Number:
Molecular Weight:
475.57

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1