
Gly-His-Lys acetate salt

Synonym(s):
Liver cell growth factor
Linear Formula:
C14H24N6O4C2H4O2
CAS Number:
Molecular Weight:
400.43
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Sterilization Method
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing